3-(3-Aminopropoxy)benzonitrile 95%
Fornitore: Apollo Scientific
Formula:
C₁₀H₁₂N₂O Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD16495891 CAS Number: 785760-03-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...