Stampa…

1,2-Dimethyl-1H-pyrrole-3-carboxylic acid 95%

Fornitore: Apollo Scientific

OR950518-1G OR950518-250MG
APOSOR950518-1GEA 199 EUR
APOSOR950518-1G APOSOR950518-250MG
1,2-Dimethyl-1H-pyrrole-3-carboxylic acid 95%
1,2-Dimethyl-1H-pyrrole-3-carboxylic acid
Formula: C₇H₉NO₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD19230065
CAS Number: 89776-57-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR