N-Benzyl-N-methylaniline 95%
Fornitore: Apollo Scientific
OR950171-1G
OR950171-250MG
OR950171-5G
APOSOR950171-1GEA
299
EUR
APOSOR950171-1G
APOSOR950171-250MG
APOSOR950171-5G
N-Benzyl-N-methylaniline 95%
N-Benzyl-N-methylaniline
Formula:
C₁₄H₁₅N Peso molecolare: 197,28 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00035788 CAS Number: 614-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...