Stampa…

1,3,3-Trimethyl-3,4-dihydroisoquinoline 95%

Fornitore: Apollo Scientific

Synonyms: 3,4-Dihydro-1,3,3-trimethylisoquinoline

OR949800-1G OR949800-250MG
APOSOR949800-1GEA 104 EUR
APOSOR949800-1G APOSOR949800-250MG
1,3,3-Trimethyl-3,4-dihydroisoquinoline 95%
1,3,3-Trimethyl-3,4-dihydroisoquinoline
Formula: C₁₂H₁₅N
Peso molecolare: 173,26 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00608546
CAS Number: 79023-51-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR