Stampa…

5-(3-Chlorophenyl)-1,3,4-oxadiazol-2-amine 95%

Fornitore: Apollo Scientific

OR946699-1G OR946699-5G
APOSOR946699-1GEA 190 EUR
APOSOR946699-1G APOSOR946699-5G
5-(3-Chlorophenyl)-1,3,4-oxadiazol-2-amine 95%
5-(3-Chlorophenyl)-1,3,4-oxadiazol-2-amine
Formula: C₈H₆ClN₃O
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01710628
CAS Number: 1673-45-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR