Stampa…

2-Chloroethyl benzoate 95%

Fornitore: Apollo Scientific

Synonyms: Benzoic Acid 2-Chloroethyl Ester

OR944810-25G OR944810-5G
APOSOR944810-25GEA 71.3 EUR
APOSOR944810-25G APOSOR944810-5G
2-Chloroethyl benzoate 95%
2-Chloroethyl benzoate
Formula: C₉H₉ClO₂
Peso molecolare: 184,62 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00045290
CAS Number: 939-55-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR