Stampa…

Quinoxaline-2-carboxamide 95%

Fornitore: Apollo Scientific

OR944667-1G OR944667-250MG
APOSOR944667-1GEA 252 EUR
APOSOR944667-1G APOSOR944667-250MG
Quinoxaline-2-carboxamide 95%
Quinoxaline-2-carboxamide
Formula: C₉H₇N₃O
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01684982
CAS Number: 5182-90-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR