Stampa…

Methyl 3,4-Dihydroxy-2-methylbenzoate 95%

Fornitore: Apollo Scientific

Synonyms: Methyl 3,4-dihydroxy-2-methylbenzoate, 3,4-Dihydroxy-2-methylbenzoic acid methyl ester

OR943092-1G OR943092-250MG
APOSOR943092-1GEA 64 EUR
APOSOR943092-1G APOSOR943092-250MG
Methyl 3,4-Dihydroxy-2-methylbenzoate 95%
Methyl 3,4-Dihydroxy-2-methylbenzoate
Formula: C₉H₁₀O₄
Peso molecolare: 182,18 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD16250128
CAS Number: 740799-82-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR