Stampa…

4-Nitro-1-naphthol 95%

Fornitore: Apollo Scientific

Synonyms: 4-Nitronaphthalen-1-ol, KAT8-IN-1, 1-Hydroxy-4-nitronaphthalene

OR942971-10G OR942971-1G OR942971-5G
APOSOR942971-10GEA 415 EUR
APOSOR942971-10G APOSOR942971-1G APOSOR942971-5G
4-Nitro-1-naphthol 95%
4-Nitro-1-naphthol
Formula: C₁₀H₇NO₃
Peso molecolare: 189,17 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD02179392
CAS Number: 605-62-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR