4-Phenyl-2-methyl-3-butyn-2-ol 95%
Fornitore: Apollo Scientific
Synonyms:
2-Methyl-4-phenylbut-3-yn-2-ol
OR942808-1G
OR942808-5G
APOSOR942808-1GEA
96
EUR
APOSOR942808-1G
APOSOR942808-5G
4-Phenyl-2-methyl-3-butyn-2-ol 95%
4-Phenyl-2-methyl-3-butyn-2-ol
Formula:
C₁₁H₁₂O Peso molecolare: 160,2100 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00041572 CAS Number: 1719-19-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...