Stampa…

3-Methylthioazetidine hydrochloride 95%

Fornitore: Apollo Scientific
Attenzione

Synonyms: Azetidine-3-thiol hydrochloride

OR942426-1G OR942426-5G
APOSOR942426-1GEA 364 EUR
APOSOR942426-1G APOSOR942426-5G
3-Methylthioazetidine hydrochloride 95%
3-Methylthioazetidine hydrochloride
Formula: C₄H₁₀ClNS
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD17015916
CAS Number: 179337-60-1

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR