Stampa…

Methyl-2-oxocyclohexanecarboxylate 90%

Fornitore: Apollo Scientific

OR941792-1G OR941792-5G
APOSOR941792-1GEA 21.5 EUR
APOSOR941792-1G APOSOR941792-5G
Methyl-2-oxocyclohexanecarboxylate 90%
Methyl-2-oxocyclohexanecarboxylate
Formula: C₈H₁₂O₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00077699
CAS Number: 41302-34-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR