2,4-Dichloro-6-methyl-1,3,5-triazine 98%
Fornitore: Apollo Scientific
OR941454-1G
OR941454-250MG
APOSOR941454-1GEA
415
EUR
APOSOR941454-1G
APOSOR941454-250MG
2,4-Dichloro-6-methyl-1,3,5-triazine 98%
2,4-Dichloro-6-methyl-1,3,5-triazine
Formula:
C₄H₃Cl₂N₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD15203622 CAS Number: 1973-04-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...