5-Bromo-N,N-diethyl-2-methoxybenzenesulphonamide 98%
Fornitore: Apollo Scientific
OR9404-1G
OR9404-5G
APOSOR9404-1GEA
139
EUR
APOSOR9404-1G
APOSOR9404-5G
5-Bromo-N,N-diethyl-2-methoxybenzenesulphonamide 98%
5-Bromo-N,N-diethyl-2-methoxybenzenesulphonamide
Formula:
C₁₁H₁₆BrNO₃S Punto di fusione: 90…94 °C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD02919424 CAS Number: 428471-30-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...