2-(3,5-Dimethoxyphenyl)propan-2-ol 95%
Fornitore: Apollo Scientific
Formula:
C₁₁H₁₆O₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD02684175 CAS Number: 39507-96-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...