Stampa…

Dichloroacetaldehyde hydrate 90%

Fornitore: Apollo Scientific

Synonyms: 2,2-Dichloro-1,1-ethanediol

OR939237-1G OR939237-25G OR939237-5G
APOSOR939237-1GEA 147 EUR
APOSOR939237-1G APOSOR939237-25G APOSOR939237-5G
Dichloroacetaldehyde hydrate 90%
Dichloroacetaldehyde hydrate
Formula: C₂H₂Cl₂O
Peso molecolare: 112,94 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00150652
CAS Number: 16086-14-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR