Stampa…

2-(2-Hydroxyphenyl)ethanol 95%

Fornitore: Apollo Scientific

Synonyms: 2-(2-Hydroxyethyl)phenol, 2-Hydroxyphenethyl alcohol

OR939175-1G OR939175-5G
APOSOR939175-1GEA 59.7 EUR
APOSOR939175-1G APOSOR939175-5G
2-(2-Hydroxyphenyl)ethanol 95%
2-(2-Hydroxyphenyl)ethanol
Formula: C₈H₁₀O₂
Peso molecolare: 138,17 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002890
CAS Number: 7768-28-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR