Stampa…

(S)-1-(Pyridin-4-yl)ethanol 95%

Fornitore: Apollo Scientific

Synonyms: (S)-alpha-Methyl-4-pyridinemethanol, (S)-1-(4-Pyridyl)ethanol, (S)-4-(1-Hydroxyethyl)pyridine

OR939111-10G OR939111-1G OR939111-5G
APOSOR939111-10GEA 452 EUR
APOSOR939111-10G APOSOR939111-1G APOSOR939111-5G
(S)-1-(Pyridin-4-yl)ethanol 95%
(S)-1-(Pyridin-4-yl)ethanol
Formula: C₇H₉NO
Peso molecolare: 123,155 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00077866
CAS Number: 54656-96-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR