Stampa…

trans-4-Propylcyclohexanol 95%

Fornitore: Apollo Scientific

OR939021-1G OR939021-250MG
APOSOR939021-1GEA 735 EUR
APOSOR939021-1G APOSOR939021-250MG
trans-4-Propylcyclohexanol 95%
trans-4-Propylcyclohexanol
Formula: C₉H₁₈O
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11114411
CAS Number: 77866-58-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR