3-(Dicyanomethylidene)indan-1-one 95%
Fornitore: Apollo Scientific
Synonyms:
2-(3-Oxo-2,3-dihydro-1H-inden-1-ylidene)malononitrile
OR938865-1G
OR938865-250MG
APOSOR938865-1GEA
148
EUR
APOSOR938865-1G
APOSOR938865-250MG
3-(Dicyanomethylidene)indan-1-one 95%
3-(Dicyanomethylidene)indan-1-one
Formula:
C₁₂H₆N₂O Peso molecolare: 194,19 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00143117 CAS Number: 1080-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...