Stampa…

6-Methoxypurine 95%

Fornitore: Apollo Scientific

OR938675-1G OR938675-5G
APOSOR938675-1GEA 24.4 EUR
APOSOR938675-1G APOSOR938675-5G
6-Methoxypurine 95%
6-Methoxypurine
Formula: C₆H₆N₄O
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00005574
CAS Number: 1074-89-1

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR