Stampa…
3-Bromo-3-methyl-2-butanone 96%
Fornitore: Apollo Scientific
OR938435-25G OR938435-5G
APOSOR938435-25GEA 458 EUR
APOSOR938435-25G APOSOR938435-5G
3-Bromo-3-methyl-2-butanone 96%
3-Bromo-3-methyl-2-butanone
Formula: C₅H₉BrO
MDL Number: MFCD06661470
CAS Number: 2648-71-7