Stampa…

2-Methoxybutyl acetate 95%

Fornitore: Apollo Scientific

Synonyms: Acetic Acid 2-Methoxybutyl Ester

OR938072-10G OR938072-1G OR938072-5G
APOSOR938072-10GEA 575 EUR
APOSOR938072-10G APOSOR938072-1G APOSOR938072-5G
2-Methoxybutyl acetate 95%
2-Methoxybutyl acetate
Formula: C₇H₁₄O₃
Peso molecolare: 146,19 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06797047
CAS Number: 1173168-18-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR