Hexanophenone 95%
Fornitore: Apollo Scientific
OR937978-100G
OR937978-25G
OR937978-5G
APOSOR937978-100GEA
119
EUR
APOSOR937978-100G
APOSOR937978-25G
APOSOR937978-5G
Hexanophenone 95%
Hexanophenone
Formula:
C₁₂H₁₆O Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00009512 CAS Number: 942-92-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...