Stampa…

N-Metil-o-toluidina 95%

Fornitore: Apollo Scientific

Synonyms: 2-(Methylamino)toluene, 2-Methyl-N-methylaniline, o-(Methylamino)toluene, N-Methyl-2-methylaniline, N,2-Dimethylaniline

OR937939-25G OR937939-5G
APOSOR937939-25GEA 270 EUR
APOSOR937939-25G APOSOR937939-5G
N-Metil-o-toluidina 95%
N-Metil-o-toluidina
Formula: C₈H₁₁N
Peso molecolare: 121,18 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00025627
CAS Number: 611-21-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR