Stampa…

(S)-2-Chloro-4-methylvaleric acid 95%

Fornitore: Apollo Scientific

Synonyms: (S)-2-Chloro-4-methylpentanoic Acid

OR937739-1G OR937739-5G
APOSOR937739-1GEA 314 EUR
APOSOR937739-1G APOSOR937739-5G
(S)-2-Chloro-4-methylvaleric acid 95%
(S)-2-Chloro-4-methylvaleric acid
Formula: C₆H₁₁ClO₂
Peso molecolare: 150,60 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00044943
CAS Number: 28659-81-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR