Stampa…

(4-Methyl-1,3-thiazol-5-yl)methanol 95%

Fornitore: Apollo Scientific

Synonyms: 4-Methyl-5-thiazolemethanol, (4-Methylthiazol-5-yl)methanol

OR937622-1G OR937622-250MG
APOSOR937622-1GEA 597 EUR
APOSOR937622-1G APOSOR937622-250MG
(4-Methyl-1,3-thiazol-5-yl)methanol 95%
(4-Methyl-1,3-thiazol-5-yl)methanol
Formula: C₅H₇NOS
Peso molecolare: 129,18 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09032911
CAS Number: 1977-06-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR