Stampa…

2-(2-Chloroethoxy)tetrahydropyran 96%

Fornitore: Apollo Scientific

Synonyms: 2-(2-Chloroethoxy)tetrahydro-2H-pyran

OR937581-25G OR937581-5G
APOSOR937581-25GEA 306 EUR
APOSOR937581-25G APOSOR937581-5G
2-(2-Chloroethoxy)tetrahydropyran 96%
2-(2-Chloroethoxy)tetrahydropyran
Formula: C₇H₁₃ClO₂
Peso molecolare: 164,6300 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00142761
CAS Number: 5631-96-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR