2-(Ethylmethylamino)pyridine 95%
Fornitore: Apollo Scientific
Formula:
C₈H₁₂N₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD04038419 CAS Number: 77200-12-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...