Stampa…

N,N-Dimethylnicotinamide 95%

Fornitore: Apollo Scientific

OR937538-10G OR937538-1G OR937538-5G
APOSOR937538-10GEA 466 EUR
APOSOR937538-10G APOSOR937538-1G APOSOR937538-5G
N,N-Dimethylnicotinamide 95%
N,N-Dimethylnicotinamide
Formula: C₈H₁₀N₂O
Peso molecolare: 150,18 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00059169
CAS Number: 6972-69-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR