Stampa…

6-Heptynenitrile 95%

Fornitore: Apollo Scientific

OR937409-10G OR937409-1G OR937409-5G
APOSOR937409-10GEA 964 EUR
APOSOR937409-10G APOSOR937409-1G APOSOR937409-5G
6-Heptynenitrile 95%
6-Heptynenitrile
Formula: C₇H₉N
Temperatura di stoccaggio: Ambiente
CAS Number: 15295-69-9

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR