Stampa…

3-Cyclohexylpropionic acid 98%

Fornitore: Apollo Scientific

Synonyms: 3-Cyclohexylpropanoic acid, 3-Cyclohexylpropionic Acid

OR937399-100G OR937399-1KG OR937399-500G
APOSOR937399-100GEA 68.4 EUR
APOSOR937399-100G APOSOR937399-1KG APOSOR937399-500G
3-Cyclohexylpropionic acid 98%
3-Cyclohexylpropionic acid
Formula: C₉H₁₆O₂
Peso molecolare: 156,22 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00001527
CAS Number: 701-97-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR