1,4-Dimethoxy-2-nitrobenzene 95%
Fornitore: Apollo Scientific
Synonyms:
2-Nitrohydroquinone dimethyl ether, Nitrodimethylhydroquinone
OR937358-25G
OR937358-5G
APOSOR937358-25GEA
306
EUR
APOSOR937358-25G
APOSOR937358-5G
1,4-Dimethoxy-2-nitrobenzene 95%
1,4-Dimethoxy-2-nitrobenzene
Formula:
C₈H₉NO₄ Peso molecolare: 183,16 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00024211 CAS Number: 89-39-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...