Stampa…

(S)-(+)-2-Methylglutaric acid 98%

Fornitore: Apollo Scientific

OR936647-1G OR936647-5G
APOSOR936647-1GEA 306 EUR
APOSOR936647-1G APOSOR936647-5G
(S)-(+)-2-Methylglutaric acid 98%
(S)-(+)-2-Methylglutaric acid
Formula: C₆H₁₀O₄
Temperatura di stoccaggio: Ambiente
CAS Number: 1115-82-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR