Stampa…

2-Nitroso-1-naftolo 98%

Fornitore: Apollo Scientific

Synonyms: 1-Hydroxy-2-nitrosonaphthalene

OR936641-25G OR936641-5G
APOSOR936641-25GEA 350 EUR
APOSOR936641-25G APOSOR936641-5G
2-Nitroso-1-naftolo 98%
2-Nitroso-1-naftolo
Formula: C₁₀H₇NO₂
Peso molecolare: 173,17 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003932
CAS Number: 132-53-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR