α,α-Diethoxytoluene 95%
Fornitore: Apollo Scientific
Formula:
C₁₁H₁₆O₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00038525 CAS Number: 774-48-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...