Stampa…

Methyl-5-chlorovalerate 95%

Fornitore: Apollo Scientific

OR936431-100G OR936431-25G OR936431-5G
APOSOR936431-100GEA 928 EUR
APOSOR936431-100G APOSOR936431-25G APOSOR936431-5G
Methyl-5-chlorovalerate 95%
Methyl-5-chlorovalerate
Formula: C₆H₁₁ClO₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00065274
CAS Number: 14273-86-0

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR