Stampa…

4'-Acetoxyacetophenone 95%

Fornitore: Apollo Scientific

OR936428-100G OR936428-25G
APOSOR936428-100GEA 30.1 EUR
APOSOR936428-100G APOSOR936428-25G
4'-Acetoxyacetophenone 95%
4'-Acetoxyacetophenone
Formula: C₁₀H₁₀O₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00017229
CAS Number: 13031-43-1

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR