3-Butylacetylacetone 98%
Fornitore: Apollo Scientific
Synonyms:
3-n-Butyl-2,4-pentanedione
OR936245-1G
OR936245-25G
OR936245-5G
APOSOR936245-1GEA
132
EUR
APOSOR936245-1G
APOSOR936245-25G
APOSOR936245-5G
3-Butylacetylacetone 98%
3-Butylacetylacetone
Formula:
C₉H₁₆O₂ Peso molecolare: 156,22 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00026235 CAS Number: 1540-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...