4-Phenyl-3-butyn-2-one 95%
Fornitore: Apollo Scientific
OR932117-10G
OR932117-1G
OR932117-5G
APOSOR932117-10GEA
887
EUR
APOSOR932117-10G
APOSOR932117-1G
APOSOR932117-5G
4-Phenyl-3-butyn-2-one 95%
4-Phenyl-3-butyn-2-one
Formula:
C₁₀H₈O Peso molecolare: 144,17 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00008776 CAS Number: 1817-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...