Stampa…

S-Ethyl thiopropionate 95%

Fornitore: Apollo Scientific

Synonyms: Thiopropionic Acid S-Ethyl Ester

OR930878-25G OR930878-5G
APOSOR930878-25GEA 299 EUR
APOSOR930878-25G APOSOR930878-5G
S-Ethyl thiopropionate 95%
S-Ethyl thiopropionate
Formula: C₅H₁₀OS
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00027016
CAS Number: 2432-42-0

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR