Stampa…

1-(2-Chloroethyl)-4-ethyl-1,4-dihydro-5H-tetrazol-5-one 95%

Fornitore: Apollo Scientific

OR929983-1G OR929983-250MG
APOSOR929983-1GEA 488 EUR
APOSOR929983-1G APOSOR929983-250MG
1-(2-Chloroethyl)-4-ethyl-1,4-dihydro-5H-tetrazol-5-one 95%
1-(2-Chloroethyl)-4-ethyl-1,4-dihydro-5H-tetrazol-5-one
Formula: C₅H₉ClN₄O
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09907794
CAS Number: 69049-03-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR