Stampa…

5-Propyl-isoxazole-3-carboxylic acid 95%

Fornitore: Apollo Scientific

OR929396-1G OR929396-5G
APOSOR929396-1GEA 190 EUR
APOSOR929396-1G APOSOR929396-5G
5-Propyl-isoxazole-3-carboxylic acid 95%
5-Propyl-isoxazole-3-carboxylic acid
Formula: C₇H₉NO₃
Peso molecolare: 155,15 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD05668697
CAS Number: 89776-75-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR