Stampa…

trans-2-Octenoic acid 95%

Fornitore: Apollo Scientific

OR929368-10G OR929368-1G OR929368-5G
APOSOR929368-10GEA 161 EUR
APOSOR929368-10G APOSOR929368-1G APOSOR929368-5G
trans-2-Octenoic acid 95%
trans-2-Octenoic acid
Formula: C₈H₁₄O₂
Peso molecolare: 142,20 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002706
CAS Number: 1871-67-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR