Stampa…

3-Pyrrolidin-1-ylpropan-1-ol 98%

Fornitore: Apollo Scientific

Synonyms: 3-(1-Pyrrolidyl)-1-propanol

OR928862-10G OR928862-1G OR928862-5G
APOSOR928862-10GEA 698 EUR
APOSOR928862-10G APOSOR928862-1G APOSOR928862-5G
3-Pyrrolidin-1-ylpropan-1-ol 98%
3-Pyrrolidin-1-ylpropan-1-ol
Formula: C₇H₁₅NO
Peso molecolare: 129,2 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01687769
CAS Number: 19748-66-4
UN: 2924
ADR: 3+8,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR