Stampa…

(2E)-3-(1-Ethyl-3,5-dimethyl-1H-pyrazol-4-yl)acrylic acid 95%

Fornitore: Apollo Scientific

Synonyms: 3-(1-Ethyl-3,5-dimethyl-1H-pyrazol-4-yl)prop-2-enoic acid

OR928429-1G OR928429-250MG
APOSOR928429-1GEA 720 EUR
APOSOR928429-1G APOSOR928429-250MG
(2E)-3-(1-Ethyl-3,5-dimethyl-1H-pyrazol-4-yl)acrylic acid 95%
(2E)-3-(1-Ethyl-3,5-dimethyl-1H-pyrazol-4-yl)acrylic acid
Formula: C₁₀H₁₄N₂O₂
Peso molecolare: 194,24 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD02090859
CAS Number: 514800-93-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR