Stampa…

2-(Morpholin-3-yl)acetic acid hydrochloride 97%

Fornitore: Apollo Scientific

OR928013-1G OR928013-250MG OR928013-500MG
APOSOR928013-1GEA 982 EUR
APOSOR928013-1G APOSOR928013-250MG APOSOR928013-500MG
2-(Morpholin-3-yl)acetic acid hydrochloride 97%
2-(Morpholin-3-yl)acetic acid hydrochloride
Formula: C₆H₁₂ClNO₃
Peso molecolare: 181,62 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11506159
CAS Number: 86967-55-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR