Stampa…

4-Butylmorpholine 95%

Fornitore: Apollo Scientific

OR926210-25G OR926210-5G
APOSOR926210-25GEA 451 EUR
APOSOR926210-25G APOSOR926210-5G
4-Butylmorpholine 95%
4-Butylmorpholine
Formula: C₈H₁₇NO
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00023401
CAS Number: 1005-67-0

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR