Stampa…

6-Chloro-3-methylpicolinic acid 98%

Fornitore: Apollo Scientific

OR926007-1G OR926007-250MG
APOSOR926007-1GEA 437 EUR
APOSOR926007-1G APOSOR926007-250MG
6-Chloro-3-methylpicolinic acid 98%
6-Chloro-3-methylpicolinic acid
Formula: C₇H₆ClNO₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD16249840
CAS Number: 1201924-32-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR