Stampa…

3-(3-Methylbutoxy)benzaldehyde 95%

Fornitore: Apollo Scientific

OR925871-1G OR925871-5G
APOSOR925871-1GEA 219 EUR
APOSOR925871-1G APOSOR925871-5G
3-(3-Methylbutoxy)benzaldehyde 95%
3-(3-Methylbutoxy)benzaldehyde
Formula: C₁₂H₁₆O₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09864622
CAS Number: 77422-25-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR